siRNA Details
| Gene Symbol: Rnf128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001254761.1 | NP_001241690.1 | 6 | 1153 | ||
| NM_023270.5 | NP_075759.3 | 6 | 1279 | |||
| GenBank mRNA | AB041548.1 | - | 6 | 1279 | ||
| AF426411.1 | - | 6 | 1093 | |||
| AK004847.1 | - | 6 | 1280 | |||
| AK018582.1 | - | 6 | 1281 | |||
| AK146266.1 | - | 6 | 1276 | |||
| AK167031.1 | - | 6 | 1265 | |||
| AY112656.1 | - | 6 | 1266 | |||
| BC010477.1 | - | 6 | 1268 | 3154433 | ||

Rnf128
Genomic Map