siRNA Details
| Gene Symbol: Rnf128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001254761.1 | NP_001241690.1 | 7 | 1303 | ||
| NM_023270.5 | NP_075759.3 | 7 | 1429 | |||
| GenBank mRNA | AB041548.1 | - | 7 | 1429 | ||
| AF426411.1 | - | 7 | 1243 | |||
| AK004847.1 | - | 7 | 1430 | |||
| AK018582.1 | - | 7 | 1431 | |||
| AK146266.1 | - | 7 | 1426 | |||
| AK167031.1 | - | 7 | 1415 | |||
| AY112656.1 | - | 7 | 1416 | |||
| BC010477.1 | - | 7 | 1418 | 3154433 | ||

Rnf128
Genomic Map