siRNA Details
| Gene Symbol: Rnf128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001254761.1 | NP_001241690.1 | 4 | 925 | ||
| NM_023270.5 | NP_075759.3 | 4 | 1051 | |||
| GenBank mRNA | AB041548.1 | - | 4 | 1051 | ||
| AF426411.1 | - | 4 | 865 | |||
| AK004847.1 | - | 4 | 1052 | |||
| AK018582.1 | - | 4 | 1053 | |||
| AK146266.1 | - | 4 | 1048 | |||
| AK167031.1 | - | 4 | 1037 | |||
| AY112656.1 | - | 4 | 1038 | |||
| BC010477.1 | - | 4 | 1040 | 3154433 | ||
| DV064262.1 | - | 4 | 688 | |||

Rnf128
Genomic Map