siRNA Details
| Gene Symbol: Lman2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025828.3 | NP_080104.2 | 3 | 541 | ||
| GenBank mRNA | AK004952.1 | - | 3 | 372 | ||
| AK014384.1 | - | 3 | 363 | |||
| AK083442.1 | - | 3 | 373 | |||
| AK129028.1 | - | 3 | 368 | |||
| AK150383.1 | - | 3 | 370 | |||
| AK166953.1 | - | 3 | 378 | |||
| AK169401.1 | - | 3 | 369 | |||
| BC049221.1 | - | 3 | 365 | 2599070 | ||
| BC055327.1 | - | 3 | 541 | 5702182 | ||

Lman2
Genomic Map