siRNA Details
| Gene Symbol: Baiap2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025833.3 | NP_080109.1 | 9 | 1093 | ||
| GenBank mRNA | AK004918.2 | - | 9 | 1094 | ||
| AK008203.1 | - | 9 | 994 | |||
| AK162158.1 | - | 1123 | ||||
| BC015459.1 | - | 9 | 871 | 3592632 | ||

Baiap2l1
Genomic Map