siRNA Details
| Gene Symbol: Baiap2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025833.3 | NP_080109.1 | 5 | 594 | ||
| GenBank mRNA | AK004918.2 | - | 5 | 595 | ||
| AK008203.1 | - | 5 | 495 | |||
| BC015459.1 | - | 5 | 372 | 3592632 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Baiap2l1| Gene Symbol: Baiap2l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025833.3 | NP_080109.1 | 5 | 594 | ||
| GenBank mRNA | AK004918.2 | - | 5 | 595 | ||
| AK008203.1 | - | 5 | 495 | |||
| BC015459.1 | - | 5 | 372 | 3592632 | ||