siRNA Details
| Gene Symbol: Plin3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025836.3 | NP_080112.1 | 5 | 478 | ||
| GenBank mRNA | AK004970.1 | - | 5 | 479 | ||
| AK009148.1 | - | 1 | 25 | |||
| AK012517.1 | - | 5 | 479 | |||
| AK049537.1 | - | 5 | 479 | |||
| AK050140.1 | - | 5 | 480 | |||
| AK132313.1 | - | 479 | ||||
| AK167214.1 | - | 5 | 477 | |||
| AK184102.1 | - | 3 | 186 | |||
| BC011116.1 | - | 5 | 474 | 4023354 | ||
| DQ257436.1 | - | 5 | 437 | |||

Plin3
Genomic Map