siRNA Details
Gene Symbol: Fam188a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024185.4 | NP_077147.2 | 6 | 711 | ||
GenBank mRNA | AK002900.1 | - | 6 | 662 | ||
AK009891.1 | - | 6 | 690 | |||
AK010050.1 | - | 6 | 687 | |||
AK049694.1 | - | 6 | 693 | |||
AK049869.1 | - | 5 | 635 | |||
AK075792.1 | - | 4 | 481 | |||
AK143250.1 | - | 6 | 683 | |||
BC002225.1 | - | 6 | 693 | |||
BC027202.1 | - | 6 | 615 | 4217391 |
