siRNA Details
Gene Symbol: Fam188a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024185.4 | NP_077147.2 | 13 | 1294 | ||
GenBank mRNA | AK002900.1 | - | 13 | 1245 | ||
AK009891.1 | - | 13 | 1273 | |||
AK010050.1 | - | 13 | 1260 | |||
AK049869.1 | - | 12 | 1218 | |||
AK075792.1 | - | 11 | 1064 | |||
AK192959.1 | - | 2 | 156 | |||
AK193641.1 | - | 2 | 140 | |||
BC002225.1 | - | 13 | 1276 | |||
BC027202.1 | - | 13 | 1198 | 4217391 |
