siRNA Details
Gene Symbol: Fam188a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024185.4 | NP_077147.2 | 11 | 1125 | ||
GenBank mRNA | AK002900.1 | - | 11 | 1076 | ||
AK009891.1 | - | 11 | 1104 | |||
AK017916.1 | - | 1 | 1700 | |||
AK049869.1 | - | 10 | 1049 | |||
AK075792.1 | - | 9 | 895 | |||
AK143250.1 | - | 11 | 1097 | |||
AK208803.1 | - | 5 | 351 | |||
BC002225.1 | - | 11 | 1107 | |||
BC027202.1 | - | 11 | 1029 | 4217391 |
