siRNA Details
Gene Symbol: Ssbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024186.5 | NP_077148.1 | 12 | 836 | ||
NM_024272.4 | NP_077234.2 | 13 | 926 | |||
GenBank mRNA | AK010220.1 | - | 12 | 778 | ||
AK013177.1 | - | 13 | 852 | |||
AK133699.1 | - | 12 | 781 | |||
AY037837.1 | - | 13 | 882 | |||
AY167988.1 | - | 13 | 836 | |||
BC002176.1 | - | 12 | 848 | 3486988 | ||
EF410127.1 | - | 9 | 808 |
