siRNA Details
| Gene Symbol: Ssbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024186.5 | NP_077148.1 | 7, 8 | 558 | ||
| NM_024272.4 | NP_077234.2 | 8, 9 | 648 | |||
| GenBank mRNA | AK010220.1 | - | 7, 8 | 500 | ||
| AK013177.1 | - | 8, 9 | 574 | |||
| AK133699.1 | - | 7, 8 | 502 | |||
| AY037837.1 | - | 8, 9 | 604 | |||
| AY167988.1 | - | 8, 9 | 558 | |||
| BC002176.1 | - | 7, 8 | 570 | 3486988 | ||
| EF410127.1 | - | 4, 5 | 530 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: XM_008760652, XM_008760648, XM_008760649, XM_008760653, FQ221531.

Ssbp2
Genomic Map