siRNA Details
| Gene Symbol: Khdc3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025890.3 | NP_080166.1 | 2 | 421 | ||
| GenBank mRNA | AB211060.1 | - | 2 | 358 | ||
| AK010377.1 | - | 2 | 358 | |||
| AK145595.1 | - | 2 | 421 | |||
| AK166731.1 | - | 2 | 371 | |||
| BC137808.1 | - | 2 | 329 | 8860827 | ||
| BC137809.1 | - | 2 | 329 | 8860828 | ||
| DQ917421.1 | - | 2 | 309 | |||
| DQ917422.1 | - | 2 | 309 | |||

Khdc3
Genomic Map