siRNA Details
| Gene Symbol: Psmd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080554.2 | NP_542121.2 | 9 | 1190 | ||
| GenBank mRNA | AK005335.1 | - | 9 | 1168 | ||
| AK011026.1 | - | 9 | 1167 | |||
| AK029067.1 | - | 7 | 2567 | |||
| AK032992.1 | - | 9 | 1168 | |||
| AK078419.1 | - | 9 | 1168 | |||
| AK145587.1 | - | 9 | 1167 | |||
| AK148284.1 | - | 1191 | ||||
| AK208538.1 | - | 1 | 14 | |||
| AK210792.1 | - | 2 | 130 | |||
| AK213478.1 | - | 1 | 14 | |||
| AK220321.1 | - | 9 | 1131 | |||
| BC010650.1 | - | 3 | 291 | |||
| BC019112.1 | - | 9 | 1201 | |||

Psmd5
Genomic Map