siRNA Details
| Gene Symbol: Uqcrc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025899.2 | NP_080175.1 | 8, 9 | 776 | ||
| GenBank mRNA | AK003975.1 | - | 8, 9 | 750 | ||
| AK004492.1 | - | 8, 9 | 751 | |||
| AK005151.1 | - | 8, 9 | 760 | |||
| AK007669.1 | - | 7, 8 | 640 | |||
| AK075856.1 | - | 8, 9 | 720 | |||
| AK077583.1 | - | 8, 9 | 725 | |||
| AK088103.1 | - | 8, 9 | 730 | |||
| AK167377.1 | - | 8, 9 | 730 | |||
| BC003423.1 | - | 8, 9 | 748 | 2609295 | ||

Uqcrc2
Genomic Map