siRNA Details
Gene Symbol: Ttc23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025905.3 | NP_080181.2 | 7 | 1282 | ||
GenBank mRNA | BC038179.1 | - | 7 | 1253 | 4485073 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ttc23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025905.3 | NP_080181.2 | 7 | 1282 | ||
GenBank mRNA | BC038179.1 | - | 7 | 1253 | 4485073 |