siRNA Details
| Gene Symbol: Oma1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025909.3 | NP_080185.1 | 8 | 1317 | ||
| GenBank mRNA | AK008020.1 | - | 8 | 1318 | ||
| AK028706.1 | - | 1 | 3170 | |||
| AK076209.1 | - | 8 | 1347 | |||
| BC016238.1 | - | 7 | 1272 | 4504835 | ||

Oma1
Genomic Map