siRNA Details
| Gene Symbol: Tbc1d2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_194334.2 | NP_919315.2 | 12 | 2722 | ||
| GenBank mRNA | AK089175.1 | - | 12 | 2688 | ||
| AK129273.1 | - | 12 | 2708 | |||
| AK156788.1 | - | 12 | 2709 | |||
| BC030847.1 | - | 2 | 132 | |||
| BC056467.1 | - | 5 | 1033 | |||
| BC108420.1 | - | 7 | 1327 | |||

Tbc1d2b
Genomic Map