siRNA Details
| Gene Symbol: Ndc80 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023294.2 | NP_075783.2 | 13 | 1555 | ||
| GenBank mRNA | AF326730.1 | - | 13 | 1568 | ||
| AK011497.1 | - | 13 | 1525 | |||
| AK011554.1 | - | 9 | 951 | |||
| AK136236.1 | - | 12 | 1374 | |||
| AK163317.1 | - | 13 | 1506 | |||
| AK169427.1 | - | 13 | 1512 | |||
| BC020131.1 | - | 13 | 1491 | 3709641 | ||

Ndc80
Genomic Map