siRNA Details
| Gene Symbol: Ola1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025942.2 | NP_080218.1 | 4 | 439 | ||
| NM_030091.1 | NP_084367.1 | 4 | 439 | |||
| GenBank mRNA | AK010817.1 | - | 4 | 355 | ||
| AK012998.1 | - | 4 | 405 | |||
| AK013055.1 | - | 4 | 440 | |||
| AK017803.1 | - | 4 | 316 | |||
| AK019142.1 | - | 4 | 325 | |||
| AK159873.1 | - | 4 | 356 | |||
| AK167458.1 | - | 4 | 355 | |||
| AK170982.1 | - | 4 | 369 | |||
| AK199282.1 | - | 3 | 292 | |||
| AK205353.1 | - | 3 | 243 | |||
| BC011318.1 | - | 4 | 339 | 3985271 | ||
| BC029207.1 | - | 4 | 332 | 4009561 | ||
| BG069761.2 | - | 5 | 577 | |||

Ola1
Genomic Map