siRNA Details
| Gene Symbol: Lsm14a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025948.2 | NP_080224.1 | 4 | 516 | ||
| GenBank mRNA | AK003611.1 | - | 2 | 76 | ||
| AK012276.1 | - | 3 | 279 | |||
| AK182421.1 | - | 2 | 183 | |||
| AK186175.1 | - | 1 | 36 | |||
| AK189673.1 | - | 3 | 251 | |||
| AK205955.1 | - | 2 | 184 | |||
| BC031521.1 | - | 4 | 516 | 2655161 | ||

Lsm14a
Genomic Map