siRNA Details
| Gene Symbol: Rps6ka6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025949.3 | NP_080225.2 | 23, 24 | 2479 | ||
| GenBank mRNA | AK012150.2 | - | 12, 13 | 1214 | ||
| AK014822.1 | - | 2, 3 | 143 | |||
| AK049349.1 | - | 21, 22 | 2230 | |||

Rps6ka6
Genomic Map