siRNA Details
| Gene Symbol: Rps6ka6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025949.3 | NP_080225.2 | 4, 5 | 587 | ||
| GenBank mRNA | AK049349.1 | - | 2, 3 | 338 | ||
| BC054113.1 | - | 2, 3 | 260 | 30015425 | ||
| CN694399.1 | - | 4, 5 | 573 | |||

Rps6ka6
Genomic Map