siRNA Details
| Gene Symbol: D16Ertd472e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252438.1 | NP_001239367.1 | 3, 4 | 1148 | ||
| NM_001252439.1 | NP_001239368.1 | 3, 4 | 915 | |||
| NM_001252440.1 | NP_001239369.1 | 3, 4 | 880 | |||
| NM_025967.4 | NP_080243.3 | 3, 4 | 948 | |||
| GenBank mRNA | AK009258.1 | - | 3, 4 | 1119 | ||
| AK044670.1 | - | 3, 4 | 949 | |||
| AW536354.1 | - | 2, 3 | 399 | |||
| BC019957.1 | - | 3, 4 | 839 | 3966397 | ||

D16Ertd472e
Genomic Map