siRNA Details
| Gene Symbol: D16Ertd472e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001252438.1 | NP_001239367.1 | 3 | 973 | ||
| NM_001252439.1 | NP_001239368.1 | 3 | 740 | |||
| NM_001252440.1 | NP_001239369.1 | 3 | 705 | |||
| NM_025967.4 | NP_080243.3 | 3 | 773 | |||
| GenBank mRNA | AK009258.1 | - | 3 | 944 | ||
| AK044670.1 | - | 3 | 774 | |||
| AW536354.1 | - | 3 | 574 | |||
| BC019957.1 | - | 3 | 664 | 3966397 | ||

D16Ertd472e
Genomic Map