siRNA Details
| Gene Symbol: Ptgr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025968.3 | NP_080244.1 | 2 | 110 | ||
| GenBank mRNA | AK010888.1 | - | 2 | 145 | ||
| AK011962.1 | - | 2 | 132 | |||
| AK035425.1 | - | 2 | 111 | |||
| AK134440.1 | - | 2 | 143 | |||
| AK166835.1 | - | 2 | 110 | |||
| BC014865.1 | - | 2 | 92 | 2648051 | ||
| CT010267.1 | - | 1 | 59 | |||

Ptgr1
Genomic Map