siRNA Details
| Gene Symbol: Bfar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177552.1 | NP_001171023.1 | 6 | 1257 | ||
| NM_025976.5 | NP_080252.1 | 8 | 1632 | |||
| GenBank mRNA | AK013874.1 | - | 8 | 1497 | ||
| AK028592.1 | - | 6 | 1096 | |||
| BC027221.1 | - | 8 | 1473 | 4196466 | ||

Bfar
Genomic Map