siRNA Details
| Gene Symbol: Ttc14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290500.1 | NP_001277429.1 | 5 | 755 | ||
| NM_001290502.1 | NP_001277431.1 | 5 | 993 | |||
| NM_025978.4 | NP_080254.2 | 5 | 755 | |||
| NM_027619.4 | NP_081895.2 | 5 | 755 | |||
| GenBank mRNA | AK012246.1 | - | 5 | 717 | ||
| AK015308.1 | - | 5 | 715 | |||
| AK016619.1 | - | 5 | 675 | |||
| AK017763.1 | - | 3 | 353 | |||
| AK031605.1 | - | 5 | 986 | |||
| AK035570.1 | - | 5 | 725 | |||
| AK041095.1 | - | 5 | 729 | |||
| AK044266.1 | - | 5 | 730 | |||
| AK134539.1 | - | 5 | 744 | |||
| AK173315.1 | - | 5 | 725 | |||
| BC021448.1 | - | 5 | 763 | |||
| BC024847.1 | - | 5 | 720 | 5355806 | ||
| BC138212.1 | - | 5 | 717 | 8861231 | ||
| BC145944.1 | - | 5 | 717 | 40131247 | ||

Ttc14
Genomic Map