siRNA Details
| Gene Symbol: Mastl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025979.4 | NP_080255.3 | 8 | 2178 | ||
| GenBank mRNA | AK006448.1 | - | 3 | 1279 | ||
| AK012597.1 | - | 3 | 1279 | |||
| AK030140.1 | - | 8 | 2073 | |||
| BC103779.1 | - | 8 | 2054 | 30480071 | ||

Mastl
Genomic Map