siRNA Details
| Gene Symbol: Ubap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290454.1 | NP_001277383.1 | 5 | 1359 | ||
| NM_023305.3 | NP_075794.3 | 6 | 1542 | |||
| GenBank mRNA | AF275549.3 | - | 6 | 1501 | ||
| AK009596.1 | - | 5 | 1318 | |||
| AK028477.1 | - | 6 | 1501 | |||
| AK028670.1 | - | 6 | 1493 | |||
| AK030370.1 | - | 6 | 1483 | |||
| AK033121.1 | - | 6 | 1494 | |||
| AK051346.1 | - | 723 | ||||
| AK076078.1 | - | 6 | 1501 | |||
| AK212879.1 | - | 2 | 122 | |||
| BC065996.1 | - | 6 | 1490 | 5689710 | ||

Ubap1
Genomic Map