siRNA Details
| Gene Symbol: Tspan31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025982.4 | NP_080258.1 | 5, 6 | 650 | ||
| GenBank mRNA | AK012457.1 | - | 5, 6 | 651 | ||
| AK012567.1 | - | 5, 6 | 653 | |||
| AK012853.1 | - | 5, 6 | 653 | |||
| AK014183.1 | - | 5, 6 | 649 | |||
| AK075962.1 | - | 5, 6 | 651 | |||
| BC023306.1 | - | 5, 6 | 627 | 4165774 | ||

Tspan31
Genomic Map