siRNA Details
| Gene Symbol: Kbtbd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025991.3 | NP_080267.1 | 2 | 550 | ||
| GenBank mRNA | AK010998.1 | - | 2 | 551 | ||
| AK014373.1 | - | 2 | 476 | |||
| BC025103.1 | - | 2 | 455 | 4481431 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kbtbd4| Gene Symbol: Kbtbd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025991.3 | NP_080267.1 | 2 | 550 | ||
| GenBank mRNA | AK010998.1 | - | 2 | 551 | ||
| AK014373.1 | - | 2 | 476 | |||
| BC025103.1 | - | 2 | 455 | 4481431 | ||