siRNA Details
| Gene Symbol: Lrrc40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289524.1 | NP_001276453.1 | 15 | 1783 | ||
| NM_001289525.1 | NP_001276454.1 | 13 | 1530 | |||
| NM_024194.6 | NP_077156.2 | 15 | 1786 | |||
| GenBank mRNA | AK011726.1 | - | 15 | 1767 | ||
| AK017814.1 | - | 15 | 1755 | |||
| AK034978.1 | - | 1766 | ||||
| AK148748.1 | - | 15 | 1746 | |||
| AK152244.1 | - | 13 | 1511 | |||
| AK163288.1 | - | 15 | 1754 | |||
| AK170052.1 | - | 15 | 1763 | |||
| BC002279.1 | - | 11 | 1175 | 3497400 | ||

Lrrc40
Genomic Map