siRNA Details
| Gene Symbol: Lrrc40 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289524.1 | NP_001276453.1 | 12 | 1387 | ||
| NM_001289525.1 | NP_001276454.1 | 10 | 1134 | |||
| NM_024194.6 | NP_077156.2 | 12 | 1390 | |||
| GenBank mRNA | AK011726.1 | - | 12 | 1371 | ||
| AK017814.1 | - | 12 | 1359 | |||
| AK034978.1 | - | 1370 | ||||
| AK148748.1 | - | 12 | 1350 | |||
| AK152244.1 | - | 10 | 1115 | |||
| AK163288.1 | - | 12 | 1358 | |||
| AK170052.1 | - | 12 | 1367 | |||
| BC002279.1 | - | 8 | 779 | 3497400 | ||

Lrrc40
Genomic Map