siRNA Details
| Gene Symbol: Lpar6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175116.4 | NP_780325.1 | 1 | 741 | ||
| GenBank mRNA | AK008952.1 | - | 1 | 617 | ||
| AK011967.1 | - | 1 | 633 | |||
| AK032901.1 | - | 1 | 738 | |||
| AK169602.1 | - | 1 | 630 | |||
| BC047931.1 | - | 1 | 617 | 5251236 | ||
| BC057175.1 | - | 1 | 604 | |||
| BC069991.1 | - | 1 | 590 | 30244233 | ||
| BC145940.1 | - | 1 | 367 | 40131243 | ||
| BC145942.1 | - | 1 | 367 | 40131245 | ||

Lpar6
Genomic Map