siRNA Details
| Gene Symbol: Spats2l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164566.1 | NP_001158038.1 | 10, 11 | 1183 | ||
| NM_144882.4 | NP_659131.2 | 12, 13 | 1369 | |||
| GenBank mRNA | AK031266.1 | - | 12, 13 | 1324 | ||
| AK032169.1 | - | 12, 13 | 1304 | |||
| BC014764.1 | - | 10, 11 | 1183 | 4217648 | ||
| BC021879.1 | - | 12, 13 | 1305 | 4216816 | ||
| BC029001.1 | - | 1, 2 | 102 | |||
| BE335142.1 | - | 2, 3 | 130 | |||
| CK624383.1 | - | 5, 6 | 476 | |||

Spats2l
Genomic Map