siRNA Details
| Gene Symbol: Prpf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026045.3 | NP_080321.2 | 3 | 280 | ||
| GenBank mRNA | AK035073.1 | - | 3 | 310 | ||
| AK135707.1 | - | 3 | 320 | |||
| AK135890.1 | - | 3 | 301 | |||
| AK139720.1 | - | 3 | 268 | |||
| BC004573.1 | - | 3 | 280 | 3586127 | ||
| BC028306.1 | - | 3 | 283 | |||
| BC058631.1 | - | 3 | 275 | 6402639 | ||

Prpf18
Genomic Map