siRNA Details
| Gene Symbol: Zfp329 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026046.3 | NP_080322.2 | 2 | 619 | ||
| GenBank mRNA | AK013271.1 | - | 2 | 610 | ||
| AK014562.2 | - | 2 | 614 | |||
| AK016779.1 | - | 1 | 436 | |||
| AK051535.1 | - | 2 | 618 | |||
| BB656010.1 | - | 2 | 618 | |||
| BC072665.1 | - | 2 | 600 | 30543571 | ||
| BC139021.1 | - | 1 | 589 | 8862043 | ||
| BC139023.1 | - | 1 | 589 | 8862045 | ||

Zfp329
Genomic Map