siRNA Details
Gene Symbol: Peli1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023324.2 | NP_075813.2 | 5 | 745 | ||
GenBank mRNA | AF302503.1 | - | 4 | 967 | ||
AK151660.1 | - | 5 | 746 | |||
AK152900.1 | - | 5 | 746 | |||
AK152997.1 | - | 5 | 745 | |||
AK153372.1 | - | 5 | 746 | |||
AK167525.1 | - | 5 | 746 | |||
BC016515.1 | - | 4 | 344 | 3600130 | ||
BC026695.1 | - | 5 | 709 |
