siRNA Details
| Gene Symbol: Peli1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_023324.2 | NP_075813.2 | 7 | 1366 | ||
| GenBank mRNA | AF302503.1 | - | 6 | 1588 | ||
| AK009945.1 | - | 1 | 175 | |||
| AK013388.1 | - | 1 | 1136 | |||
| AK029586.1 | - | 6 | 3128 | |||
| AK076458.1 | - | 1 | 88 | |||
| AK151660.1 | - | 7 | 1367 | |||
| AK152900.1 | - | 7 | 1367 | |||
| AK152997.1 | - | 7 | 1366 | |||
| AK153372.1 | - | 7 | 1367 | |||
| AK167525.1 | - | 7 | 1367 | |||
| BC016515.1 | - | 6 | 965 | 3600130 | ||

Peli1
Genomic Map