siRNA Details
| Gene Symbol: Fam69a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026062.4 | NP_080338.1 | 5 | 754 | ||
| GenBank mRNA | AK008504.1 | - | 5 | 698 | ||
| AK013580.1 | - | 5 | 718 | |||
| AK038811.1 | - | 5 | 712 | |||
| AK048979.1 | - | 5 | 711 | |||
| AK050305.1 | - | 5 | 711 | |||
| AK053273.1 | - | 4 | 559 | |||
| BC055842.1 | - | 5 | 686 | |||
| BC061013.1 | - | 5 | 706 | |||
| BC095935.1 | - | 5 | 688 | |||
| BC116683.1 | - | 5 | 699 | 40106565 | ||

Fam69a
Genomic Map