siRNA Details
| Gene Symbol: Pigc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039045.1 | NP_001034134.1 | 2 | 666 | ||
| NM_026078.2 | NP_080354.1 | 2 | 679 | |||
| GenBank mRNA | AK008524.1 | - | 2 | 631 | ||
| AK014096.1 | - | 2 | 641 | |||
| AK016394.1 | - | 2 | 678 | |||
| AK152660.1 | - | 2 | 657 | |||
| AK155971.1 | - | 1 | 1588 | |||
| AK170153.1 | - | 2 | 634 | |||
| BC006938.1 | - | 2 | 599 | 2649047 | ||

Pigc
Genomic Map