siRNA Details
| Gene Symbol: Dock7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001290636.1 | NP_001277565.1 | 30 | 3839 | ||
| NM_026082.5 | NP_080358.3 | 29 | 3749 | |||
| GenBank mRNA | BC086672.1 | - | 10 | 1257 | 30647593 | |
| BC094420.1 | - | 17 | 2035 | |||
| BM964053.1 | - | 4 | 501 | |||

Dock7
Genomic Map