siRNA Details
Gene Symbol: Cltc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003908.1 | NP_001003908.1 | 6 | 1043 | ||
GenBank mRNA | AK122193.1 | - | 6 | 1047 | ||
AK156869.1 | - | 6 | 1062 | |||
AK158438.1 | - | 6 | 1029 | |||
AK171732.1 | - | 6 | 1045 | |||
BC079897.1 | - | 6 | 1043 | 30546407 |
