siRNA Details
| Gene Symbol: Zc3h13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026083.2 | NP_080359.2 | 4 | 559 | ||
| GenBank mRNA | AK014200.1 | - | 4 | 339 | ||
| AK078225.1 | - | 4 | 550 | |||
| BC138264.1 | - | 4 | 508 | 8861283 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zc3h13| Gene Symbol: Zc3h13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026083.2 | NP_080359.2 | 4 | 559 | ||
| GenBank mRNA | AK014200.1 | - | 4 | 339 | ||
| AK078225.1 | - | 4 | 550 | |||
| BC138264.1 | - | 4 | 508 | 8861283 | ||