siRNA Details
| Gene Symbol: Zc2hc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173181.3 | NP_775273.1 | 8 | 821 | ||
| GenBank mRNA | AK014202.1 | - | 8 | 757 | ||
| AK083941.1 | - | 8 | 765 | |||
| AK085187.1 | - | 8 | 759 | |||
| BC044809.1 | - | 8 | 743 | |||
| BC087731.1 | - | 785 | 6850296 | |||

Zc2hc1a
Genomic Map