siRNA Details
| Gene Symbol: Zc2hc1a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_173181.3 | NP_775273.1 | 7 | 747 | ||
| GenBank mRNA | AK014202.1 | - | 7 | 683 | ||
| AK083941.1 | - | 7 | 691 | |||
| AK085187.1 | - | 7 | 685 | |||
| AK170609.1 | - | 7 | 688 | |||
| BC087731.1 | - | 711 | 6850296 | |||

Zc2hc1a
Genomic Map