siRNA Details
| Gene Symbol: 1700022I11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026088.3 | NP_080364.1 | 2 | 283 | ||
| GenBank mRNA | AK006242.1 | - | 2 | 284 | ||
| AK007056.1 | - | 2 | 243 | |||
| AK133216.1 | - | 2 | 271 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

1700022I11Rik| Gene Symbol: 1700022I11Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026088.3 | NP_080364.1 | 2 | 283 | ||
| GenBank mRNA | AK006242.1 | - | 2 | 284 | ||
| AK007056.1 | - | 2 | 243 | |||
| AK133216.1 | - | 2 | 271 | |||