siRNA Details
| Gene Symbol: Herc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026101.4 | NP_080377.2 | 9 | 1468 | ||
| NM_030114.2 | NP_084390.1 | 9 | 1468 | |||
| GenBank mRNA | AK019546.1 | - | 9 | 1464 | ||
| AK143111.1 | - | 9 | 1342 | |||
| AK145735.1 | - | 4 | 416 | |||
| AK147723.1 | - | 9 | 1355 | |||
| BC043082.1 | - | 9 | 1280 | 6417111 | ||
| BC060033.1 | - | 7 | 1053 | 6478123 | ||

Herc4
Genomic Map