siRNA Details
| Gene Symbol: Tmco5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026104.4 | NP_080380.1 | 12 | 935 | ||
| GenBank mRNA | AK007078.1 | - | 12 | 919 | ||
| BC049568.1 | - | 12 | 899 | 6742928 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmco5| Gene Symbol: Tmco5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_026104.4 | NP_080380.1 | 12 | 935 | ||
| GenBank mRNA | AK007078.1 | - | 12 | 919 | ||
| BC049568.1 | - | 12 | 899 | 6742928 | ||